Assay Design
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BC030742.1 |
- |
1 |
1145 |
|
Chr. 18 - 62976215 - 62977444 [+] on Build GRCh38 |
84 |
GenBankmRNA |
AF150170.1 |
- |
1 |
1667 |
|
Chr. 18 - 62975626 - 62977426 [+] on Build GRCh38 |
84 |
GenBankmRNA |
BM680082.1 |
- |
1 |
|
|
Chr. 18 - 62976718 - 62977446 [-] on Build GRCh38 |
84 |
GenBankmRNA |
BQ004395.1 |
- |
1 |
|
|
Chr. 18 - 62976765 - 62977446 [-] on Build GRCh38 |
84 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
zinc finger CCHC-type containing 2 |
ZCCHC2 |
C18orf49 |
-388.5kb
|
Sense |
Chr. - 62523410 - 62587709 on Build GRCh38 |
Downstream |
BCL2, apoptosis regulator |
BCL2 |
Bcl-2, PPP1R50 |
145.9kb
|
Anti-sense |
Chr. - 63123346 - 63319778 on Build GRCh38 |
View Full Product Details