Assay Design
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CR595802.1 |
- |
1 |
552 |
|
Chr. 19 - 19630065 - 19631110 [-] on Build GRCh38 |
78 |
GenBankmRNA |
AL534214.3 |
- |
1 |
552 |
|
Chr. 19 - 19630145 - 19631110 [-] on Build GRCh38 |
78 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
GEM interacting protein |
GMIP |
ARHGAP46 |
|
Sense |
Chr. - 19629473 - 19643667 on Build GRCh38 |
Downstream |
ATPase 13A1 |
ATP13A1 |
ATP13A, CGI-152 |
14.1kb
|
Sense |
Chr. - 19645198 - 19663694 on Build GRCh38 |
View Full Product Details