Assay Design
Gene Transcripts
- Gene Symbol
- STIM1
- Entrez Gene ID
-
6786
- Gene Name
- stromal interaction molecule 1
- Gene Aliases
-
D11S4896E, GOK, IMD10, STRMK, TAM, TAM1
- Chromosome Location
-
Chr.11: 3855703 - 4093210 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK057599.1 |
- |
1 - 1 |
1646 |
60 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB342455.1 |
- |
1 |
|
|
Chr. 11 - 3980066 - 3980610 [-] on Build GRCh38 |
60 |
GenBankmRNA |
BQ022900.1 |
- |
1 |
|
|
Chr. 11 - 3980197 - 3980714 [-] on Build GRCh38 |
60 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 4687 |
MIR4687 |
|
-122.7kb
|
Sense |
Chr. - 3856062 - 3856141 on Build GRCh38 |
Downstream |
ribonucleotide reductase catalytic subunit M1 |
RRM1 |
R1, RIR1, RR1 |
114.0kb
|
Sense |
Chr. - 4094685 - 4138925 on Build GRCh38 |
View Full Product Details