Assay Design
Gene Transcripts
- Gene Symbol
- LIPJ
- Entrez Gene ID
-
142910
- Gene Name
- lipase family member J
- Gene Aliases
-
LIPL1, bA425M17.2
- Chromosome Location
-
Chr.10: 88583914 - 88623161 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK090891.1 |
- |
1 - 1 |
1645 |
103 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB301764.1 |
- |
1 |
|
|
Chr. 10 - 88585364 - 88585908 [-] on Build GRCh38 |
103 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
renalase, FAD dependent amine oxidase |
RNLS |
C10orf59, RENALASE |
-60.0
|
Anti-sense |
Chr. - 88176458 - 88583860 on Build GRCh38 |
Downstream |
lipase F, gastric type |
LIPF |
GL, HGL, HLAL |
78.5kb
|
Sense |
Chr. - 88664389 - 88678815 on Build GRCh38 |
View Full Product Details