Assay Design
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AK091382.1 |
- |
1 |
1747 |
|
Chr. 4 - 8493911 - 8496275 [+] on Build GRCh38 |
73 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
tRNA methyltransferase 44 homolog (S. cerevisiae) |
TRMT44 |
C4orf23, METTL19, TRM44 |
|
Sense |
Chr. - 8440774 - 8512531 on Build GRCh38 |
Downstream |
G protein-coupled receptor 78 |
GPR78 |
|
84.0kb
|
Sense |
Chr. - 8580237 - 8590023 on Build GRCh38 |
View Full Product Details