Assay Design
Gene Transcripts
- Gene Symbol
- LOC101243545
- Entrez Gene ID
-
101243545
- Gene Name
- uncharacterized LOC101243545
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 161426427 - 161448242 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_102265.2 |
|
3 - 4 |
1444 |
94 |
GenBank mRNA |
AK094885.1 |
- |
3 - 4 |
1446 |
94 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
serine palmitoyltransferase small subunit B |
SPTSSB |
ADMP, C3orf57, SSSPTB |
-70.7kb
|
Anti-sense |
Chr. - 161344784 - 161373123 on Build GRCh38 |
Downstream |
long intergenic non-protein coding RNA 1192 |
LINC01192 |
CT64 |
1.7mb
|
Anti-sense |
Chr. - 163177243 - 163303301 on Build GRCh38 |
View Full Product Details