Assay Design



Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK095239.1 - 1 2349 Chr. 10 - 4980066 - 4983280 [+] on Build GRCh38 72


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream aldo-keto reductase family 1 member C1 AKR1C1 2-ALPHA-HSD, 20-ALPHA-HSD, C9, DD1, DD1/DD2, DDH, DDH1, H-37, HAKRC, HBAB, MBAB -2.1kb Sense Chr. - 4963262 - 4979967 on Build GRCh38
Downstream aldo-keto reductase family 1, member C2 AKR1C2 AKR1C-pseudo, BABP, DD, DD-2, DD/BABP, DD2, DDH2, HAKRD, HBAB, MCDR2, SRXY8, TDD 4.5kb Anti-sense Chr. - 4987775 - 5018033 on Build GRCh38

View Full Product Details