Assay Design



Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AW880203.1 - 1 148 Chr. 18 - 63307171 - 63307556 [-] on Build GRCh38 78
GenBankmRNA AK024118.1 - 1 2250 Chr. 18 - 63306858 - 63309668 [-] on Build GRCh38 78
GenBankmRNA BX089092.1 - 1 190 Chr. 18 - 63306886 - 63307608 [-] on Build GRCh38 78
GenBankmRNA AW880202.1 - 1 149 Chr. 18 - 63307160 - 63307556 [-] on Build GRCh38 78
GenBankmRNA CD685722.1 - 1 399 Chr. 18 - 63307180 - 63307808 [-] on Build GRCh38 78


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream PH domain and leucine rich repeat protein phosphatase 1 PHLPP1 PHLPP, PLEKHE1, PPM3A, SCOP -326.4kb Anti-sense Chr. - 62715439 - 62980443 on Build GRCh38
Downstream 3-ketodihydrosphingosine reductase KDSR DHSR, FVT1, SDR35C1 18.1kb Sense Chr. - 63327736 - 63367273 on Build GRCh38

View Full Product Details