Assay Design
Gene Transcripts
- Gene Symbol
- RPL13
- Entrez Gene ID
-
6137
- Gene Name
- ribosomal protein L13
- Gene Aliases
-
BBC1, D16S444E, D16S44E, L13
- Chromosome Location
-
Chr.16: 89560657 - 89566829 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB326293.1 |
- |
1 |
|
|
Chr. 16 - 89566290 - 89566829 [-] on Build GRCh38 |
88 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
small nucleolar RNA, C/D box 68 |
SNORD68 |
HBII-202 |
-2.9kb
|
Sense |
Chr. - 89561430 - 89561501 on Build GRCh38 |
Downstream |
copine 7 |
CPNE7 |
|
8.9kb
|
Sense |
Chr. - 89575768 - 89597246 on Build GRCh38 |
View Full Product Details