Assay Design



Gene Transcripts

Gene Symbol
HHIPL1
Entrez Gene ID
84439
Gene Name
HHIP like 1
Gene Aliases
KIAA1822, UNQ9245
Chromosome Location
Chr.14: 99604476 - 99680569 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006720277.3 XP_006720340.1 1 - 2 139 71
XM_011537236.2 XP_011535538.1 1 - 2 139 71
XM_017021708.1 XP_016877197.1 1 - 2 139 71
GenBank mRNA BC039687.1 - 1 - 2 59 71


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream coiled-coil domain containing 85C CCDC85C -166.0 Anti-sense Chr. - 99511266 - 99604390 on Build GRCh38
Downstream cytochrome P450 family 46 subfamily A member 1 CYP46A1 CP46, CYP46 58.5kb Sense Chr. - 99684262 - 99727321 on Build GRCh38

View Full Product Details