Assay Design
Gene Transcripts
- Gene Symbol
- TMEM59
- Entrez Gene ID
-
9528
- Gene Name
- transmembrane protein 59
- Gene Aliases
-
C1orf8, DCF1, HSPC001, PRO195, UNQ169
- Chromosome Location
-
Chr.1: 54026681 - 54053573 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK129688.1 |
- |
1 - 1 |
450 |
77 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
low density lipoprotein receptor class A domain containing 1 |
LDLRAD1 |
|
-14.9kb
|
Sense |
Chr. - 54007298 - 54018186 on Build GRCh38 |
Downstream |
transcription elongation factor A N-terminal and central domain containing 2 |
TCEANC2 |
C1orf83 |
18.7kb
|
Anti-sense |
Chr. - 54053572 - 54112519 on Build GRCh38 |
View Full Product Details