Assay Design



Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK123657.1 - 1 1657 Chr. 11 - 17381406 - 17383531 [+] on Build GRCh38 64
GenBankmRNA DA507423.1 - 1 Chr. 11 - 17382695 - 17383232 [-] on Build GRCh38 64
GenBankmRNA DB317799.1 - 1 Chr. 11 - 17383008 - 17383531 [-] on Build GRCh38 64


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream natural killer cell cytotoxicity receptor 3 ligand 1 NCR3LG1 B7-H6, B7H6, DKFZp686O24166 Sense Chr. - 17351762 - 17377321 on Build GRCh38
Downstream ATP binding cassette subfamily C member 8 ABCC8 ABC36, HHF1, HI, HRINS, MRP8, PHHI, SUR, SUR1, SUR1delta2, TNDM2 9.4kb Anti-sense Chr. - 17392885 - 17476870 on Build GRCh38

View Full Product Details