Assay Design
Gene Transcripts
- Gene Symbol
- RBM6
- Entrez Gene ID
-
10180
- Gene Name
- RNA binding motif protein 6
- Gene Aliases
-
3G2, DEF-3, DEF3, HLC-11, NY-LU-12, g16
- Chromosome Location
-
Chr.3: 49940038 - 50077252 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK124030.1 |
- |
1 - 1 |
57 |
101 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DA173932.1 |
- |
1 |
254 |
|
Chr. 3 - 50067493 - 50068067 [+] on Build GRCh38 |
101 |
GenBankmRNA |
DB098123.1 |
- |
1 |
548 |
|
Chr. 3 - 50067199 - 50067790 [+] on Build GRCh38 |
101 |
GenBankmRNA |
DA958615.1 |
- |
1 |
57 |
|
Chr. 3 - 50067689 - 50068247 [+] on Build GRCh38 |
101 |
GenBankmRNA |
BM908060.1 |
- |
1 |
157 |
|
Chr. 3 - 50067601 - 50068550 [+] on Build GRCh38 |
101 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
MON1 homolog A, secretory trafficking associated |
MON1A |
SAND1 |
-137.3kb
|
Anti-sense |
Chr. - 49908869 - 49930012 on Build GRCh38 |
Downstream |
RNA binding motif protein 5 |
RBM5 |
G15, H37, LUCA15, RMB5 |
18.6kb
|
Sense |
Chr. - 50088908 - 50118964 on Build GRCh38 |
View Full Product Details