Assay Design
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB348710.1 |
- |
1 |
|
|
Chr. 18 - 37246153 - 37246688 [-] on Build GRCh38 |
95 |
GenBankmRNA |
AK126277.1 |
- |
1 |
2454 |
|
Chr. 18 - 37243767 - 37246688 [+] on Build GRCh38 |
95 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105372069 |
LOC105372069 |
|
-319.9kb
|
Anti-sense |
Chr. - 36901945 - 36923814 on Build GRCh38 |
Downstream |
uncharacterized LOC105372068 |
LOC105372068 |
|
27.8kb
|
Sense |
Chr. - 37274460 - 37276400 on Build GRCh38 |
View Full Product Details