Assay Design
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AK130551.1 |
- |
1 |
445 |
|
Chr. 12 - 101916892 - 101917694 [+] on Build GRCh38 |
85 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
N-acetylglucosamine-1-phosphate transferase alpha and beta subunits |
GNPTAB |
GNPTA, ICD |
-86.0kb
|
Anti-sense |
Chr. - 101745497 - 101830867 on Build GRCh38 |
Downstream |
coiled-coil domain containing 53 |
CCDC53 |
CGI-116 |
95.1kb
|
Anti-sense |
Chr. - 102012840 - 102062124 on Build GRCh38 |
View Full Product Details