Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003718.4 | NP_003709.3 | 14 - 14 | 6248 | 88 |
NM_031267.3 | NP_112557.2 | 14 - 14 | 6068 | 88 | |
XM_017012750.1 | XP_016868239.1 | 15 - 15 | 7197 | 88 | |
XM_017012751.1 | XP_016868240.1 | 15 - 15 | 7017 | 88 | |
GenBank mRNA | AK130653.1 | - | 1 - 1 | 442 | 88 |
BM727913.1 | - | 1 - 1 | 333 | 88 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 265 | LINC00265 | NCRNA00265 | -301.3kb | Sense | Chr. - 39733568 - 39794623 on Build GRCh38 |
Downstream | M-phase specific PLK1 interacting protein | MPLKIP | ABHS, C7orf11, ORF20, TTD4 | 35.6kb | Anti-sense | Chr. - 40132743 - 40134652 on Build GRCh38 |