Assay Design
Gene Transcripts
- Gene Symbol
- LOC100505564
- Entrez Gene ID
-
100505564
- Gene Name
- uncharacterized LOC100505564
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK055529.1 |
- |
1 - 2 |
313 |
80 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DA559788.1 |
- |
1 - 2 |
313 |
|
Chr. 16 - 10940719 - 10941427 [+] on Build GRCh38 |
80 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
trans-golgi network vesicle protein 23 homolog A |
TVP23A |
FAM18A, YDR084C |
-121.9kb
|
Anti-sense |
Chr. - 10758034 - 10818902 on Build GRCh38 |
Downstream |
C-type lectin domain family 16 member A |
CLEC16A |
Gop-1, KIAA0350 |
1.5kb
|
Sense |
Chr. - 10944488 - 11193272 on Build GRCh38 |
View Full Product Details