Assay Design
Gene Transcripts
- Gene Symbol
- LRAT
- Entrez Gene ID
-
9227
- Gene Name
- lecithin retinol acyltransferase (phosphatidylcholine--retinol O-acyltransferase)
- Gene Aliases
-
LCA14
- Chromosome Location
-
Chr.4: 154740841 - 154753119 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK056234.1 |
- |
2 - 3 |
277 |
97 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DA702216.1 |
- |
2 - 3 |
277 |
|
Chr. 4 - 154626945 - 154698639 [+] on Build GRCh38 |
97 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
fibrinogen gamma chain |
FGG |
|
-23.6kb
|
Anti-sense |
Chr. - 154604134 - 154612808 on Build GRCh38 |
Downstream |
RNA binding motif protein 46 |
RBM46 |
CT68 |
81.4kb
|
Sense |
Chr. - 154781250 - 154828813 on Build GRCh38 |
View Full Product Details