Assay Design
Gene Transcripts
- Gene Symbol
- RASEF
- Entrez Gene ID
-
158158
- Gene Name
- RAS and EF-hand domain containing
- Gene Aliases
-
RAB45
- Chromosome Location
-
Chr.9: 82979585 - 83063177 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DN914144.1 |
- |
1 |
585 |
|
Chr. 9 - 82980398 - 82981073 [-] on Build GRCh38 |
68 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
SPATA31 subfamily D member 1 |
SPATA31D1 |
FAM75D1 |
-984.3kb
|
Anti-sense |
Chr. - 81987404 - 81995256 on Build GRCh38 |
Downstream |
uncharacterized LOC102723989 |
LOC102723989 |
|
236.9kb
|
Anti-sense |
Chr. - 83219288 - 83270833 on Build GRCh38 |
View Full Product Details