Assay Design
Gene Transcripts
- Gene Symbol
- GTDC1
- Entrez Gene ID
-
79712
- Gene Name
- glycosyltransferase like domain containing 1
- Gene Aliases
-
Hmat-Xa, mat-Xa
- Chromosome Location
-
Chr.2: 143945787 - 144333092 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF086278.1 |
- |
1 - 1 |
253 |
106 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
W69585.1 |
- |
|
255 |
|
Chr. - - [-] on Build |
107 |
GenBankmRNA |
W69586.1 |
- |
|
|
|
Chr. - - [-] on Build |
106 |
GenBankmRNA |
BX109452.1 |
- |
1 |
253 |
|
Chr. 2 - 143938068 - 143938606 [-] on Build GRCh38 |
106 |
GenBankmRNA |
BM664094.1 |
- |
1 |
|
|
Chr. 2 - 143938068 - 143938467 [+] on Build GRCh38 |
106 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC101928386 |
LOC101928386 |
|
|
Anti-sense |
Chr. - 143937067 - 143964156 on Build GRCh38 |
View Full Product Details