Assay Design



Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK096121.1 - 2 645 Chr. 3 - 10017491 - 10021842 [-] on Build GRCh38 96


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream cell death-inducing DFFA-like effector c pseudogene CIDECP CICE Sense Chr. - 10017553 - 10026136 on Build GRCh38
Downstream Fanconi anemia complementation group D2 FANCD2 FA-D2, FA4, FACD, FAD, FAD2, FANCD 6.3kb Anti-sense Chr. - 10026387 - 10101937 on Build GRCh38

View Full Product Details