Assay Design



Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AL117488.1 - 1 1247 Chr. Y - 21405662 - 21408270 [+] on Build GRCh38 70


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream proline rich, Y-linked PRORY CYorf17 Anti-sense Chr. - 21381674 - 21454622 on Build GRCh38
Downstream RNA binding motif protein, Y-linked, family 1, member B RBMY1B 103.5kb Sense Chr. - 21511338 - 21527212 on Build GRCh38

View Full Product Details