Assay Design
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AL133102.1 |
- |
1 |
1956 |
|
Chr. 15 - 83138591 - 83141393 [-] on Build GRCh38 |
75 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
transmembrane 6 superfamily member 1 |
TM6SF1 |
|
-576.0
|
Anti-sense |
Chr. - 83107572 - 83145403 on Build GRCh38 |
Downstream |
basonuclin 1 |
BNC1 |
BNC, BSN1, HsT19447 |
114.5kb
|
Sense |
Chr. - 83255884 - 83284716 on Build GRCh38 |
View Full Product Details