Assay Design
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AL832590.1 |
- |
2 |
4651 |
|
Chr. 18 - 13514520 - 13522888 [+] on Build GRCh38 |
86 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 5190 |
MIR5190 |
|
-59.7kb
|
Sense |
Chr. - 13459947 - 13460026 on Build GRCh38 |
Downstream |
microRNA 4526 |
MIR4526 |
mir-4526 |
88.2kb
|
Sense |
Chr. - 13611114 - 13611200 on Build GRCh38 |
View Full Product Details