Assay Design
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CA313510.1 |
- |
1 |
270 |
|
Chr. 3 - 43083471 - 43083855 [+] on Build GRCh38 |
81 |
GenBankmRNA |
AL832594.1 |
- |
1 |
3349 |
|
Chr. 3 - 43080403 - 43084427 [+] on Build GRCh38 |
81 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
family with sequence similarity 198 member A |
FAM198A |
C3orf41 |
-22.7kb
|
Sense |
Chr. - 42979267 - 43057715 on Build GRCh38 |
Downstream |
SNF related kinase |
SNRK |
HSNFRK |
202.1kb
|
Sense |
Chr. - 43286512 - 43351143 on Build GRCh38 |
View Full Product Details