Assay Design
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BM919709.1 |
- |
|
64 |
|
Chr. - - [-] on Build |
77 |
GenBankmRNA |
BC040916.1 |
- |
1 |
46 |
|
Chr. 6 - 4135426 - 4146053 [+] on Build GRCh38 |
77 |
GenBankmRNA |
DB474960.1 |
- |
1 |
|
|
Chr. 6 - 4135055 - 4135545 [-] on Build GRCh38 |
77 |
GenBankmRNA |
DB267419.1 |
- |
1 |
|
|
Chr. 6 - 4135248 - 4135635 [-] on Build GRCh38 |
77 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
chromosome 6 open reading frame 201 |
C6orf201 |
dJ1013A10.5 |
-4.7kb
|
Sense |
Chr. - 4079197 - 4130788 on Build GRCh38 |
Downstream |
uncharacterized LOC100507506 |
LOC100507506 |
|
527.0
|
Sense |
Chr. - 4136072 - 4157385 on Build GRCh38 |
View Full Product Details