Assay Design
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AJ227871.1 |
- |
1 |
55 |
|
Chr. 5 - 175503619 - 175504004 [+] on Build GRCh38 |
88 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
sideroflexin 1 |
SFXN1 |
TCC |
|
Sense |
Chr. - 175478498 - 175529742 on Build GRCh38 |
Downstream |
histamine receptor H2 |
HRH2 |
H2R |
153.8kb
|
Sense |
Chr. - 175657762 - 175710756 on Build GRCh38 |
View Full Product Details