Assay Design
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BX118687.1 |
- |
2 |
|
|
Chr. 11 - 77866412 - 77870091 [-] on Build GRCh38 |
107 |
GenBankmRNA |
AK095141.1 |
- |
|
853 |
|
Chr. - - [-] on Build |
107 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
remodeling and spacing factor 1 |
RSF1 |
HBXAP, RSF-1, XAP8, p325 |
-45.5kb
|
Sense |
Chr. - 77660009 - 77872232 on Build GRCh38 |
Downstream |
integrator complex subunit 4 |
INTS4 |
INT4, MST093 |
12.0kb
|
Sense |
Chr. - 77874491 - 77994849 on Build GRCh38 |
View Full Product Details