Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000181.3 | NP_000172.2 | 8 - 9 | 1522 | 136 |
NM_001284290.1 | NP_001271219.1 | 6 - 7 | 1084 | 136 | |
NM_001293104.1 | NP_001280033.1 | 7 - 8 | 1337 | 136 | |
NM_001293105.1 | NP_001280034.1 | 6 - 7 | 1194 | 136 | |
XM_005250297.3 | XP_005250354.1 | 7 - 8 | 1364 | 136 | |
XM_011516113.1 | XP_011514415.1 | 7 - 8 | 1266 | 136 | |
XM_011516114.1 | XP_011514416.1 | 7 - 8 | 1194 | 136 | |
XM_017012091.1 | XP_016867580.1 | 6 - 7 | 1113 | 136 | |
XM_017012092.1 | XP_016867581.1 | 6 - 7 | 1100 | 136 | |
XM_017012093.1 | XP_016867582.1 | 6 - 7 | 1041 | 136 | |
GenBank mRNA | AK223406.1 | - | 8 - 9 | 1421 | 136 |
AK300223.1 | - | 8 - 9 | 1311 | 136 | |
AK303819.1 | - | 6 - 7 | 981 | 136 | |
AK312752.1 | - | 8 - 9 | 1438 | 136 | |
BC014142.2 | - | 8 - 9 | 1392 | 136 | |
CD014092.1 | - | 7 - 8 | 1219 | 136 | |
CD014093.1 | - | 6 - 7 | 1077 | 136 | |
M15182.1 | - | 8 - 9 | 1417 | 136 |