Assay Design



Gene Transcripts

Gene Symbol
INHBB
Entrez Gene ID
3625
Gene Name
inhibin beta B subunit
Gene Aliases
-
Chromosome Location
Chr.2: 120346143 - 120351807 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002193.2 NP_002184.2 2 - 2 1495 81
GenBank mRNA BC030029.2 - 2 - 2 1495 81
M13437.1 - 2 - 2 1288 81
M31682.1 - 2 - 2 1388 81

View Full Product Details