Assay Design



Gene Transcripts

Gene Symbol
BMP15
Entrez Gene ID
9210
Gene Name
bone morphogenetic protein 15
Gene Aliases
GDF9B, ODG2, POF4
Chromosome Location
Chr.X: 50910735 - 50916641 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005448.2 NP_005439.2 2 - 2 993 79
GenBank mRNA BC069155.1 - 2 - 2 944 79
BC117264.1 - 2 - 2 993 79
BC117266.1 - 2 - 2 993 79

View Full Product Details