Assay Design



Gene Transcripts

Gene Symbol
FOXP3
Entrez Gene ID
50943
Gene Name
forkhead box P3
Gene Aliases
AIID, DIETER, IPEX, JM2, PIDX, XPID
Chromosome Location
Chr.X: 49250436 - 49266505 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114377.1 NP_001107849.1 2 - 3 399 99
XM_011543916.2 XP_011542218.1 3 - 4 1929 99
XM_017029566.1 XP_016885055.1 3 - 4 1929 99
XM_017029567.1 XP_016885056.1 2 - 3 324 99
GenBank mRNA AJ005891.1 - 2 - 3 166 99
AK299988.1 - 2 - 3 283 99
BC143785.1 - 1 - 2 241 99
DQ010327.1 - 1 - 2 211 99

View Full Product Details