Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079520.1 | NP_001072988.1 | 4 - 4 | 913 | 236 |
NM_016651.5 | NP_057735.2 | 4 - 4 | 1024 | 236 | |
NR_046093.1 | 4 - 4 | 522 | 236 | ||
NR_046095.1 | 4 - 4 | 830 | 236 | ||
XM_006720167.3 | XP_006720230.1 | 4 - 4 | 961 | 236 | |
GenBank mRNA | AF251079.2 | - | 1 - 1 | 129 | 236 |
AK299879.1 | - | 4 - 4 | 830 | 236 | |
AK316164.1 | - | 4 - 4 | 620 | 236 | |
AK316169.1 | - | 2 - 2 | 236 | 236 | |
AK316237.1 | - | 4 - 4 | 522 | 236 | |
AK316423.1 | - | 3 - 3 | 389 | 236 | |
BX161433.1 | - | 4 - 4 | 477 | 236 | |
DR157190.1 | - | 3 - 3 | 291 | 236 |