Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000147.4 | NP_000138.2 | 7 - 8 | 1348 | 96 |
XM_005245821.2 | XP_005245878.1 | 7 - 8 | 1485 | 96 | |
XM_011541167.2 | XP_011539469.1 | 7 - 8 | 933 | 96 | |
XM_017000905.1 | XP_016856394.1 | 7 - 8 | 1030 | 96 | |
GenBank mRNA | AB074175.1 | - | 7 - 8 | 1269 | 96 |
AK092914.1 | - | 8 - 9 | 1203 | 96 | |
AK095443.1 | - | 8 - 9 | 1220 | 96 | |
AK095455.1 | - | 8 - 9 | 1234 | 96 | |
AK225468.1 | - | 7 - 8 | 1269 | 96 | |
AK301302.1 | - | 9 - 10 | 1198 | 96 | |
AK314649.1 | - | 7 - 8 | 1269 | 96 | |
BC017338.2 | - | 7 - 8 | 1271 | 96 | |
M29877.1 | - | 7 - 8 | 1268 | 96 |