Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164458.1 | NP_001157930.1 | 7 - 8 | 919 | 96 |
NM_001164459.1 | NP_001157931.1 | 7 - 8 | 857 | 96 | |
GenBank mRNA | AB039791.1 | - | 7 - 8 | 857 | 96 |
BM674900.1 | - | 1 - 2 | 96 |