Assay Design



Gene Transcripts

Gene Symbol
SKA3
Entrez Gene ID
221150
Gene Name
spindle and kinetochore associated complex subunit 3
Gene Aliases
C13orf3, RAMA1
Chromosome Location
Chr.13: 21153595 - 21176602 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166017.1 NP_001159489.1 7 - 8 1239 117
GenBank mRNA AK090584.1 - 7 - 8 1184 117

View Full Product Details