Product Details

Assay Reference Genome
Location

Chr.22:38122448 on build GRCh38
Cytoband
22q13.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
8398
Gene Symbol
PLA2G6
Gene Name
phospholipase A2 group VI
Gene Aliases
CaI-PLA2, GVI, INAD1, IPLA2-VIA, NBIA2, NBIA2A, NBIA2B, PARK14, PLA2, PNPLA9, iPLA2, iPLA2beta
Location
Chr.22:38111495-38192099 on build GRCh38
Assay Gene Location
Within Intron 18
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
PLA2G6 NM_001004426.1 NP_001004426.1
NM_001199562.1 NP_001186491.1
NM_003560.2 NP_003551.2
XM_005261764.2 XP_005261821.1
XM_005261765.1 XP_005261822.1
XM_005261766.1 XP_005261823.1
XM_006724332.3 XP_006724395.1
XM_011530422.1 XP_011528724.1
XM_011530423.1 XP_011528725.1
XM_011530424.1 XP_011528726.1
XM_011530425.1 XP_011528727.1
XM_011530426.2 XP_011528728.1
XM_017028981.1 XP_016884470.1
XM_017028982.1 XP_016884471.1
XM_017028983.1 XP_016884472.1
XM_017028984.1 XP_016884473.1
XM_017028985.1 XP_016884474.1
XM_017028986.1 XP_016884475.1
AF064594.1 AAC97486.1
AF102988.1 AAD41722.1
AF102989.1 AAD41723.1
AK096521.1
AK128862.1
AK290825.1
AK291212.1
AK308308.1
AL080187.1 CAB45768.2
BC036742.1 AAH36742.2
BC051904.1 AAH51904.1
CR456543.1 CAG30429.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv471198 Chr22:38019714 - 38149612 on Build GRCh38 Loss BAIAP2L2 POLR2F SLC16A8 PLA2G6 PICK1
nsv508732 Chr22:38096355 - 38180191 on Build GRCh38 Insertion BAIAP2L2 PLA2G6
nsv834194 Chr22:38063495 - 38240504 on Build GRCh38 Loss BAIAP2L2 SNORA92 SLC16A8 MAFF PLA2G6 TMEM184B PICK1
nsv834193 Chr22:37997329 - 38172090 on Build GRCh38 Loss BAIAP2L2 POLR2F SLC16A8 PLA2G6 PICK1

View Full Product Details