Product Details

Assay Reference Genome
Location

Chr.X:14840241 on build GRCh38
Cytoband
Xp22.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
2187
Gene Symbol
FANCB
Gene Name
Fanconi anemia complementation group B
Gene Aliases
FA2, FAAP90, FAAP95, FAB, FACB
Location
Chr.X:14690863-14873255 on build GRCh38
Assay Gene Location
Within Intron 11
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
FANCB XM_011545470.2 XP_011543772.1
XM_017029356.1 XP_016884845.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv6812 ChrX:14836320 - 14868456 on Build GRCh38 Deletion FANCB
nsv9934 ChrX:14730738 - 15173654 on Build GRCh38 Loss GLRA2 MOSPD2 FANCB

View Full Product Details