Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166386.3 | NP_001159858.1 | 2 - 3 | 276 | 84 |
NM_001166387.3 | NP_001159859.1 | 2 - 3 | 184 | 84 | |
GenBank mRNA | AK314168.1 | - | 2 - 3 | 276 | 84 |
CF123053.1 | - | 2 - 3 | 181 | 84 |