Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080410.2 | NP_001073879.2 | 5 - 6 | 1802 | 66 |
XM_005264158.3 | XP_005264215.1 | 3 - 4 | 721 | 66 | |
XM_006711950.2 | XP_006712013.1 | 10 - 11 | 2786 | 66 | |
XM_011532562.1 | XP_011530864.1 | 6 - 7 | 2234 | 66 | |
XM_011532563.2 | XP_011530865.1 | 4 - 5 | 1159 | 66 | |
XM_017003421.1 | XP_016858910.1 | 7 - 8 | 4883 | 66 | |
XM_017003422.1 | XP_016858911.1 | 10 - 11 | 4528 | 66 | |
XM_017003423.1 | XP_016858912.1 | 10 - 11 | 3909 | 66 | |
GenBank mRNA | AB075820.1 | - | 5 - 6 | 1640 | 66 |
AK127503.1 | - | 2 - 3 | 283 | 66 | |
BC112202.1 | - | 4 - 5 | 710 | 66 |