Assay Design



Gene Transcripts

Gene Symbol
SHH
Entrez Gene ID
6469
Gene Name
sonic hedgehog
Gene Aliases
HHG1, HLP3, HPE3, MCOPCB5, SMMCI, TPT, TPTPS
Chromosome Location
Chr.7: 155799984 - 155812273 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000193.3 NP_000184.1 1 - 2 448 99
GenBank mRNA BX452210.1 - 1 - 2 353 99
L38518.1 - 1 - 2 448 99

View Full Product Details