Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000913.5 | NP_000904.1 | 1 - 2 | 275 | 61 |
GenBank mRNA | AF285093.1 | - | 1 - 2 | 275 | 61 |
AK308203.1 | - | 1 - 2 | 275 | 61 | |
CN267465.1 | - | 1 - 2 | 239 | 61 | |
L40949.1 | - | 1 - 2 | 222 | 61 |