Assay Design



Gene Transcripts

Gene Symbol
OR6C68
Entrez Gene ID
403284
Gene Name
olfactory receptor family 6 subfamily C member 68
Gene Aliases
-
Chromosome Location
Chr.12: 55492378 - 55493316 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005519.2 NP_001005519.2 1 - 1 435 77

View Full Product Details