Assay Design



Gene Transcripts

Gene Symbol
TEX40
Entrez Gene ID
25858
Gene Name
testis expressed 40
Gene Aliases
C11orf20
Chromosome Location
Chr.11: 64300391 - 64304767 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039496.1 NP_001034585.1 2 - 3 374 98
GenBank mRNA AL117564.1 - 2 - 3 377 98

Gene Symbol
KCNK4-TEX40
Entrez Gene ID
106780802
Gene Name
KCNK4-TEX40 readthrough
Gene Aliases
-
Chromosome Location
Chr.11: 64291722 - 64304769 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_133662.1 8 - 9 2361 98

View Full Product Details