Assay Design
Gene Transcripts
- Gene Symbol
- HYAL1
- Entrez Gene ID
-
3373
- Gene Name
- hyaluronoglucosaminidase 1
- Gene Aliases
-
HYAL-1, LUCA1, MPS9, NAT6
- Chromosome Location
-
Chr.3: 50299889 - 50312951 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_153285.2 |
NP_695017.1 |
1 - 2 |
134 |
84 |
GenBank mRNA |
AF502907.1 |
- |
1 - 2 |
106 |
84 |
View Full Product Details