Assay Design



Gene Transcripts

Gene Symbol
FAM53A
Entrez Gene ID
152877
Gene Name
family with sequence similarity 53 member A
Gene Aliases
DNTNP
Chromosome Location
Chr.4: 1574055 - 1684313 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013622.3 NP_001013644.1 1 - 2 39 100
GenBank mRNA BC070112.1 - 1 - 2 39 100

View Full Product Details