Assay Design



Gene Transcripts

Gene Symbol
GNG4
Entrez Gene ID
2786
Gene Name
G protein subunit gamma 4
Gene Aliases
-
Chromosome Location
Chr.1: 235547685 - 235650754 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098721.1 NP_001092191.1 1 - 2 207 83
GenBank mRNA DA230013.1 - 1 - 2 65 83

View Full Product Details