Assay Design



Gene Transcripts

Gene Symbol
TMEM91
Entrez Gene ID
641649
Gene Name
transmembrane protein 91
Gene Aliases
DSPC3, IFITMD6
Chromosome Location
Chr.19: 41363966 - 41384083 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098825.1 NP_001092295.1 1 - 2 129 76
GenBank mRNA BM726200.1 - 1 - 2 96 76
R01772.1 - - 128 76

View Full Product Details