Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002493.4 | NP_002484.1 | 1 - 2 | 307 | 116 |
NM_182739.2 | NP_877416.1 | 1 - 2 | 307 | 116 | |
GenBank mRNA | AF035840.1 | - | 1 - 2 | 251 | 116 |
AF067167.1 | - | 1 - 2 | 280 | 116 | |
AK289702.1 | - | 1 - 2 | 261 | 116 | |
BC009801.1 | - | 1 - 2 | 286 | 116 | |
BC016708.1 | - | 1 - 2 | 321 | 116 | |
BC029247.1 | - | 1 - 2 | 252 | 116 | |
BI669356.1 | - | 1 - 2 | 286 | 116 | |
BP208846.1 | - | 1 - 2 | 305 | 116 | |
CB140469.1 | - | 1 - 2 | 307 | 117 | |
CR456920.1 | - | 1 - 2 | 183 | 116 |