Assay Design



Gene Transcripts

Gene Symbol
ASNS
Entrez Gene ID
440
Gene Name
asparagine synthetase (glutamine-hydrolyzing)
Gene Aliases
ASNSD, TS11
Chromosome Location
Chr.7: 97852117 - 97872542 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183356.3 NP_899199.2 1 - 2 194 87
GenBank mRNA DA160359.1 - 1 - 2 105 87
DA661624.1 - 1 - 2 129 87

View Full Product Details